Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040610.2 | NP_001035700.1 | 16 | 2073 | ||
NM_001322844.1 | NP_001309773.1 | 17 | 2141 | |||
NM_001322845.1 | NP_001309774.1 | 18 | 2468 | |||
NM_024580.5 | NP_078856.4 | 18 | 2226 | |||
NR_136410.1 | 18 | 2226 | ||||
XM_011522000.1 | XP_011520302.1 | 16 | 1897 | |||
GenBank mRNA | AK023181.1 | - | 12 | 1378 | ||
AK056656.1 | - | 18 | 2156 | |||
AK300348.1 | - | 18 | 2136 | |||
BX538332.1 | - | 16 | 2004 |