Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100411.2 | NP_001093881.1 | 15 | 2875 | ||
NM_024581.5 | NP_078857.5 | 16 | 3413 | |||
GenBank mRNA | AK024004.1 | - | 9 | 1217 | ||
AK127744.1 | - | 12 | 1715 | |||
AL136767.1 | - | 5 | 528 | |||
AY131226.1 | - | 12 | 1689 | |||
BC009055.1 | - | 8 | 1005 | |||
BC060769.1 | - | 16 | 3413 | 30337859 | ||
BX538186.1 | - | 15 | 2855 |