Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193528.1 | NP_001180457.1 | 3, 4 | 341 | ||
NM_024583.4 | NP_078859.2 | 3, 4 | 432 | |||
XM_005246853.2 | XP_005246910.1 | 3, 4 | 837 | |||
XM_005246854.4 | XP_005246911.1 | 3, 4 | 1997 | |||
XM_005246855.2 | XP_005246912.1 | 3, 4 | 885 | |||
XM_005246856.2 | XP_005246913.1 | 3, 4 | 841 | |||
XM_005246857.2 | XP_005246914.1 | 3, 4 | 541 | |||
XM_011511839.2 | XP_011510141.1 | 2, 3 | 241 | |||
XM_017004909.1 | XP_016860398.1 | 3, 4 | 1429 | |||
XM_017004910.1 | XP_016860399.1 | 3, 4 | 907 | |||
GenBank mRNA | AK057912.1 | - | 3, 4 | 410 | ||
AK295357.1 | - | 3, 4 | 325 | |||
BC119684.1 | - | 3, 4 | 383 | 40116568 | ||
BC119685.1 | - | 3, 4 | 383 | 40116570 | ||
BG492502.1 | - | 3, 4 | 425 | |||
BM725063.1 | - | 2, 3 | 423 | |||
DA760451.1 | - | 3, 4 | 432 |