Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000758.3 | NP_000749.2 | 1 | 169 | ||
GenBank mRNA | AF510855.1 | - | 1 | 86 | ||
AY720441.1 | - | 1 | 146 | |||
BC108724.1 | - | 1 | 156 | 3886571 | ||
BC113924.1 | - | 1 | 160 | 40084800 | ||
BC113999.1 | - | 1 | 160 | 40084798 | ||
KP271009.1 | - | 1 | 86 | |||
M10663.1 | - | 1 | 145 | |||
M11220.1 | - | 1 | 169 | |||
M11734.1 | - | 1 | 152 |