Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142964.1 | NP_001136436.1 | 2 | 3539 | ||
XM_011530373.2 | XP_011528675.1 | 3 | 3447 | |||
XM_017028939.1 | XP_016884428.1 | 4 | 3578 | |||
GenBank mRNA | AK027237.1 | - | 1 | 452 | ||
AK131457.1 | - | 4 | 3373 | |||
BC007210.1 | - | 1 | 534 | 3344580 | ||
BC035403.1 | - | 2 | 3539 |