Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024590.3 | NP_078866.3 | 1 | 1195 | ||
XM_005263212.4 | XP_005263269.1 | 1 | 1153 | |||
XM_017008592.1 | XP_016864081.1 | 2 | 1235 | |||
XM_017008593.1 | XP_016864082.1 | 1 | 1493 | |||
XM_017008594.1 | XP_016864083.1 | 1 | 1153 | |||
XM_017008595.1 | XP_016864084.1 | 1 | 1153 | |||
XM_017008596.1 | XP_016864085.1 | 1 | 1153 | |||
XM_017008597.1 | XP_016864086.1 | 1 | 1153 | |||
XM_017008598.1 | XP_016864087.1 | 1 | 1153 | |||
XM_017008599.1 | XP_016864088.1 | 2 | 1235 | |||
GenBank mRNA | AK307532.1 | - | 2 | 756 | ||
AM049401.1 | - | 1 | 469 | |||
AY358647.1 | - | 1 | 1197 | |||
AY875938.1 | - | 1 | 307 | |||
BC033156.1 | - | 1 | 99 | |||
BC089445.1 | - | 1 | 565 | |||
BC132879.1 | - | 1 | 395 | 40146901 | ||
BC132881.1 | - | 1 | 395 | 40146903 | ||
BC144265.1 | - | 1 | 395 | 9052785 | ||
BC151213.1 | - | 1 | 1361 |