Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005498.3 | NP_001005498.2 | 16 | 2113 | ||
NM_024599.5 | NP_078875.4 | 17 | 2275 | |||
XM_005257669.3 | XP_005257726.1 | 17 | 2636 | |||
XM_005257670.1 | XP_005257727.1 | 17 | 2145 | |||
XM_011525249.2 | XP_011523551.1 | 17 | 3592 | |||
XM_011525250.2 | XP_011523552.1 | 18 | 2302 | |||
XM_011525251.2 | XP_011523553.1 | 17 | 2119 | |||
XM_017025079.1 | XP_016880568.1 | 17 | 2549 | |||
GenBank mRNA | AK025994.1 | - | 12 | 1299 | ||
AK074034.1 | - | 15 | 2580 | |||
AK225684.1 | - | 12 | 1252 | |||
AK292135.1 | - | 17 | 2268 | |||
AK292166.1 | - | 16 | 2068 | |||
AL833680.1 | - | 11 | 1168 | |||
AY500247.1 | - | 16 | 1964 | |||
BC016034.1 | - | 17 | 2255 | 3910543 | ||
BC035829.1 | - | 12 | 1361 |