Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024605.3 | NP_078881.3 | 9 | 873 | ||
XM_005263215.3 | XP_005263272.1 | 9 | 1109 | |||
XM_017008602.1 | XP_016864091.1 | 10 | 1145 | |||
GenBank mRNA | AB050785.1 | - | 9 | 873 | ||
AK304458.1 | - | 1 | 56 | |||
BC011920.2 | - | 5 | 403 | |||
BC109029.2 | - | 9 | 863 | |||
BC109030.1 | - | 9 | 862 | |||
BC126899.1 | - | 9 | 869 | |||
BC128055.1 | - | 9 | 873 | 40027832 | ||
BX647727.1 | - | 3 | 6003 |