Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024605.3 | NP_078881.3 | 7, 8 | 707 | ||
XM_005263215.3 | XP_005263272.1 | 7, 8 | 943 | |||
XM_017008602.1 | XP_016864091.1 | 8, 9 | 979 | |||
GenBank mRNA | AB050785.1 | - | 7, 8 | 707 | ||
BC011920.2 | - | 3, 4 | 237 | |||
BC109029.2 | - | 7, 8 | 697 | |||
BC109030.1 | - | 7, 8 | 696 | |||
BC126899.1 | - | 7, 8 | 703 | |||
BC128055.1 | - | 7, 8 | 707 | 40027832 | ||
BX647727.1 | - | 1, 2 | 5837 |