Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178055.1 | NP_001171526.1 | 7 | 589 | ||
NM_001178056.1 | NP_001171527.1 | 6 | 565 | |||
NM_024615.3 | NP_078891.2 | 6 | 565 | |||
XM_005248596.4 | XP_005248653.1 | 7 | 945 | |||
XM_011543631.2 | XP_011541933.1 | 7 | 478 | |||
XM_011543632.2 | XP_011541934.2 | 7 | 525 | |||
XM_011543633.2 | XP_011541935.2 | 7 | 525 | |||
XM_011543634.2 | XP_011541936.1 | 8 | 1074 | |||
XM_011543635.2 | XP_011541937.1 | 8 | 595 | |||
XM_011543636.2 | XP_011541938.1 | 7 | 459 | |||
XM_011543640.2 | XP_011541942.1 | 6 | 501 | |||
XM_011543641.2 | XP_011541943.1 | 7 | 478 | |||
XM_011543642.2 | XP_011541944.1 | 5 | 465 | |||
XM_011543643.2 | XP_011541945.1 | 6 | 906 | |||
XM_017009850.1 | XP_016865339.1 | 6 | 1025 | |||
XM_017009851.1 | XP_016865340.1 | 5 | 465 | |||
GenBank mRNA | AK289369.1 | - | 6 | 548 | ||
AK298680.1 | - | 5 | 428 | |||
AK314466.1 | - | 7 | 589 | |||
AL834477.1 | - | 6 | 524 | |||
BC037386.1 | - | 4 | 423 | 4830820 | ||
BC075801.1 | - | 6 | 524 | 5226828 | ||
BC105789.1 | - | 7 | 593 | |||
DC429180.1 | - | 6 | 537 |