Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282143.1 | NP_001269072.1 | 5 | 1002 | ||
NM_001282144.1 | NP_001269073.1 | 5 | 1410 | |||
NM_001282358.1 | NP_001269287.1 | 5 | 934 | |||
NM_024618.3 | NP_078894.2 | 5 | 1029 | |||
XM_005271669.1 | XP_005271726.1 | 5 | 1253 | |||
XM_005271670.1 | XP_005271727.1 | 4 | 906 | |||
XM_005271671.1 | XP_005271728.1 | 4 | 1080 | |||
XM_005271672.1 | XP_005271729.1 | 4 | 903 | |||
XM_006718904.1 | XP_006718967.1 | 5 | 1287 | |||
XM_011542980.1 | XP_011541282.1 | 5 | 917 | |||
XM_017018294.1 | XP_016873783.1 | 5 | 1287 | |||
GenBank mRNA | AB094095.1 | - | 5 | 1013 | ||
AK056454.1 | - | 4 | 727 | |||
AK095247.1 | - | 3 | 214 | |||
AK291716.1 | - | 5 | 996 | |||
AK303002.1 | - | 4 | 903 | |||
AY245437.1 | - | 4 | 772 | |||
BC013199.1 | - | 1 | 133 | |||
BC110890.1 | - | 5 | 1375 | 4937963 | ||
BX647705.1 | - | 5 | 987 | |||
EF452237.1 | - | 4 | 772 |