Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024619.3 | NP_078895.2 | 1 | 148 | ||
NR_046408.1 | 1 | 148 | ||||
GenBank mRNA | AK022233.1 | - | 1 | 115 | ||
AK293523.1 | - | 1 | 132 | |||
AK301958.1 | - | 1 | 132 | |||
AK315373.1 | - | 1 | 132 | |||
AL136631.1 | - | 1 | 119 | |||
AY360465.1 | - | 1 | 115 | |||
BC007611.1 | - | 1 | 124 | 3351601 | ||
BC014408.1 | - | 1 | 102 | 4652087 | ||
CR533461.1 | - | 1 | 98 |