Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FN3KRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024619.3 | NP_078895.2 | 1 | 168 | ||
NR_046408.1 | 1 | 168 | ||||
GenBank mRNA | AK022233.1 | - | 1 | 135 | ||
AK293523.1 | - | 1 | 152 | |||
AK301958.1 | - | 1 | 152 | |||
AK315373.1 | - | 1 | 152 | |||
AL136631.1 | - | 1 | 139 | |||
AY360465.1 | - | 1 | 135 | |||
BC007611.1 | - | 1 | 144 | 3351601 | ||
BC014408.1 | - | 1 | 122 | 4652087 | ||
CR533461.1 | - | 1 | 118 | |||
DA019669.1 | - | 1 | 169 |