Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000759.3 | NP_000750.1 | 5 | 802 | ||
NM_001178147.1 | NP_001171618.1 | 4 | 685 | |||
NM_172219.2 | NP_757373.1 | 5 | 793 | |||
NM_172220.2 | NP_757374.2 | 4 | 694 | |||
NR_033662.1 | 4 | 969 | ||||
GenBank mRNA | AK290247.1 | - | 4 | 953 | ||
AK298116.1 | - | 3 | 845 | |||
BC033245.1 | - | 4 | 863 | 5434958 | ||
CD013926.1 | - | 4 | 588 | |||
X03438.1 | - | 5 | 719 | |||
X03655.1 | - | 5 | 709 |