Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024627.5 | NP_078903.3 | 3 | 694 | ||
GenBank mRNA | AK024778.1 | - | 1 | 70 | ||
AK225088.1 | - | 1 | 70 | |||
AK291262.1 | - | 3 | 662 | |||
AL832509.1 | - | 3 | 583 | |||
BC011679.2 | - | 1 | 201 | 4303449 | ||
BX640785.1 | - | 2 | 783 | |||
BX640969.1 | - | 3 | 663 | |||
BX640998.1 | - | 3 | 663 |