Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024629.3 | NP_078905.2 | 8, 9 | 861 | ||
NR_104593.1 | 8, 9 | 861 | ||||
XM_005263218.4 | XP_005263275.2 | 8, 9 | 896 | |||
GenBank mRNA | AF469667.1 | - | 8, 9 | 864 | ||
AF516710.1 | - | 9, 10 | 1278 | |||
AK027121.1 | - | 6, 7 | 671 | |||
AK292297.1 | - | 8, 9 | 807 | |||
BC031520.1 | - | 3, 4 | 209 | |||
BC067802.1 | - | 8, 9 | 801 | |||
BC107744.1 | - | 3, 4 | 306 | |||
BC131556.1 | - | 8, 9 | 861 | 40108113 | ||
BC141854.1 | - | 8, 9 | 859 | 8860370 | ||
CR457376.1 | - | 1, 2 | 65 | |||
DQ907910.1 | - | 8, 9 | 791 | |||
DQ907911.1 | - | 8, 9 | 791 |