Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205315.1 | NP_001192244.1 | 4 | 893 | ||
NM_024636.3 | NP_078912.2 | 3 | 794 | |||
GenBank mRNA | AF423422.1 | - | 4 | 776 | ||
AF423423.1 | - | 4 | 776 | |||
AK026806.1 | - | 3 | 773 | |||
AK307644.1 | - | 3 | 772 |