Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024637.4 | NP_078913.3 | 4 | 1178 | ||
GenBank mRNA | AF289575.1 | - | 3 | 973 | ||
AF316113.1 | - | 4 | 1021 | |||
AK022178.1 | - | 3 | 817 | |||
AK092367.1 | - | 3 | 1804 | |||
AK225890.1 | - | 3 | 954 | |||
AK301591.1 | - | 2 | 616 | |||
AL833824.1 | - | 3 | 823 | |||
BC012976.2 | - | 4 | 997 | 3636329 |