Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024641.3 | NP_078917.2 | 4 | 868 | ||
XM_005267147.3 | XP_005267204.1 | 4 | 1261 | |||
GenBank mRNA | AJ577574.1 | - | 3 | 733 | ||
AK022900.1 | - | 3 | 374 | |||
AK291940.1 | - | 4 | 799 | |||
AY372528.1 | - | 3 | 688 | |||
BC137014.1 | - | 3 | 737 | 9021007 | ||
BC137016.1 | - | 3 | 737 | 9021009 | ||
BC146671.1 | - | 4 | 803 | 40148201 | ||
BC150199.1 | - | 4 | 803 | 40148811 | ||
BX537398.1 | - | 4 | 799 | |||
BX640869.1 | - | 4 | 829 | |||
CR936827.1 | - | 4 | 803 |