Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024646.2 | NP_078922.1 | 11 | 1942 | ||
XM_006710898.3 | XP_006710961.1 | 11 | 1829 | |||
GenBank mRNA | AB051517.1 | - | 11 | 1892 | ||
AK023518.1 | - | 5 | 399 | |||
AK056645.1 | - | 9 | 886 | |||
AK095643.1 | - | 10 | 1582 | |||
AK296460.1 | - | 9 | 1737 | |||
BC029832.1 | - | 9 | 956 | |||
BX647643.1 | - | 9 | 1115 | |||
DQ646393.1 | - | 12 | 2015 |