Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024656.3 | NP_078932.2 | 5 | 899 | ||
XM_005260080.4 | XP_005260137.1 | 5 | 545 | |||
XM_011528297.2 | XP_011526599.1 | 5 | 537 | |||
XM_011528298.2 | XP_011526600.1 | 5 | 638 | |||
XM_011528299.2 | XP_011526601.1 | 2 | 290 | |||
GenBank mRNA | AK074941.1 | - | 5 | 903 | ||
AK075541.1 | - | 5 | 791 | |||
BC071684.1 | - | 1 | 52 | |||
BC108308.1 | - | 5 | 854 | 5138787 |