Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256964.1 | NP_001243893.1 | 2 | 420 | ||
NM_001256965.2 | NP_001243894.1 | 2 | 299 | |||
NM_001256966.2 | NP_001243895.1 | 2 | 299 | |||
NM_001256967.2 | NP_001243896.1 | 2 | 294 | |||
NM_001256968.2 | NP_001243897.1 | 2 | 191 | |||
NM_001256969.2 | NP_001243898.1 | 2 | 186 | |||
NM_024661.4 | NP_078937.3 | 2 | 312 | |||
XM_011534113.2 | XP_011532415.1 | 3 | 747 | |||
GenBank mRNA | AK022498.1 | - | 2 | 191 | ||
AL533758.3 | - | 2 | 279 | |||
AW957929.1 | - | 2 | 196 | |||
BC011993.1 | - | 1 | 302 | 3857480 | ||
BG336520.1 | - | 2 | 390 | |||
BI755881.1 | - | 2 | 404 | |||
BM810621.1 | - | 2 | 322 | |||
BP307614.1 | - | 2 | 312 | |||
BQ050243.1 | - | 2 | 234 | |||
BQ427785.1 | - | 1 | 196 |