Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077447.2 | NP_001070915.1 | 3 | 513 | ||
NM_001287506.1 | NP_001274435.1 | 3 | 537 | |||
NM_001287507.1 | NP_001274436.1 | 3 | 754 | |||
NM_001287508.1 | NP_001274437.1 | 3 | 609 | |||
NM_001287509.1 | NP_001274438.1 | 3 | 468 | |||
NM_001287510.1 | NP_001274439.1 | 2 | 1131 | |||
NM_024664.3 | NP_078940.2 | 3 | 958 | |||
GenBank mRNA | AK021900.1 | - | 3 | 892 | ||
BC062586.1 | - | 2 | 1081 | |||
BC104938.1 | - | 3 | 958 | 8143941 | ||
BC106064.1 | - | 3 | 908 | 3913530 | ||
BC112015.1 | - | 3 | 958 | 8327483 | ||
BP211065.1 | - | 3 | 464 | |||
CB998485.1 | - | 3 | 421 |