Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL1XR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321193.1 | NP_001308122.1 | 11, 12 | 1314 | ||
NM_001321194.1 | NP_001308123.1 | 12, 13 | 1494 | |||
NM_001321195.1 | NP_001308124.1 | 10, 11 | 1211 | |||
NM_024665.5 | NP_078941.2 | 11, 12 | 1375 | |||
XM_005247775.2 | XP_005247832.1 | 10, 11 | 1330 | |||
XM_006713746.1 | XP_006713809.1 | 12, 13 | 1384 | |||
XM_011513141.1 | XP_011511443.1 | 12, 13 | 1318 | |||
XM_011513142.2 | XP_011511444.1 | 11, 12 | 1299 | |||
XM_011513143.2 | XP_011511445.1 | 10, 11 | 1225 | |||
XM_017007185.1 | XP_016862674.1 | 11, 12 | 1312 | |||
GenBank mRNA | AF268193.1 | - | 11, 12 | 1208 | ||
AF314544.1 | - | 10, 11 | 1174 | |||
AK022956.1 | - | 11, 12 | 1274 | |||
AK296946.1 | - | 9, 10 | 956 | |||
BC113421.1 | - | 10, 11 | 1117 | 8322473 | ||
BX648935.1 | - | 11, 12 | 1309 |