Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAGAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271885.1 | NP_001258814.1 | 5 | 512 | ||
NM_001271886.1 | NP_001258815.1 | 5 | 773 | |||
NM_024666.4 | NP_078942.3 | 5 | 628 | |||
XM_011522020.1 | XP_011520322.1 | 5 | 878 | |||
GenBank mRNA | AK021568.1 | - | 5 | 532 | ||
AK294402.1 | - | 5 | 509 | |||
AL136715.1 | - | 5 | 538 | |||
BC001975.1 | - | 2 | 149 | |||
BC047026.1 | - | 5 | 551 | 5534878 | ||
BC058886.1 | - | 5 | 550 | 6503238 | ||
CR533540.1 | - | 5 | 523 | |||
DC315715.1 | - | 5 | 512 |