Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024667.2 | NP_078943.1 | 2 | 319 | ||
XM_005253622.3 | XP_005253679.1 | 3 | 3034 | |||
XM_006719602.3 | XP_006719665.1 | 3 | 355 | |||
XM_011538743.2 | XP_011537045.1 | 3 | 460 | |||
XM_011538745.2 | XP_011537047.1 | 2 | 2850 | |||
XM_017019970.1 | XP_016875459.1 | 3 | 464 | |||
GenBank mRNA | AK022812.1 | - | 2 | 319 | ||
BC005882.2 | - | 2 | 257 | 3533384 |