Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024669.2 | NP_078945.2 | 10 | 1448 | ||
XM_011543646.1 | XP_011541948.1 | 4 | 762 | |||
XM_017009852.1 | XP_016865341.1 | 10 | 1360 | |||
XM_017009853.1 | XP_016865342.1 | 9 | 1332 | |||
XM_017009854.1 | XP_016865343.1 | 7 | 2682 | |||
GenBank mRNA | AK021857.1 | - | 2 | 501 | ||
AK123389.1 | - | 10 | 1448 | |||
BC106914.2 | - | 2 | 668 | 40032109 | ||
BC106915.2 | - | 2 | 668 | 40032110 |