Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030581.3 | NP_085058.3 | 18, 19 | 1971 | ||
XM_017023667.1 | XP_016879156.1 | 17, 18 | 1832 | |||
XM_017023668.1 | XP_016879157.1 | 16, 17 | 1608 | |||
GenBank mRNA | AB067510.1 | - | 13, 14 | 1444 | ||
AF370390.1 | - | 6, 7 | 903 | |||
AK022332.1 | - | 1, 2 | 403 | |||
AK091316.1 | - | 6, 7 | 751 | |||
AL832078.1 | - | 3, 4 | 379 | |||
BC014887.2 | - | 12, 13 | 1363 |