Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282684.1 | NP_001269613.1 | 5 | 608 | ||
NM_001282685.1 | NP_001269614.1 | 5 | 608 | |||
NM_001282686.1 | NP_001269615.1 | 5 | 608 | |||
NM_024681.3 | NP_078957.2 | 5 | 608 | |||
XM_005261741.3 | XP_005261798.2 | 5 | 619 | |||
XM_005261742.2 | XP_005261799.2 | 5 | 619 | |||
XM_005261743.2 | XP_005261800.2 | 5 | 619 | |||
XM_005261744.2 | XP_005261801.1 | 5 | 619 | |||
XM_011530374.2 | XP_011528676.1 | 5 | 619 | |||
XM_011530377.2 | XP_011528679.1 | 5 | 619 | |||
GenBank mRNA | AK022304.1 | - | 5 | 584 | ||
AK308720.1 | - | 5 | 857 | |||
BC025403.1 | - | 5 | 607 | 4824153 | ||
BC031038.1 | - | 5 | 591 | 5277700 | ||
BG832241.1 | - | 5 | 590 | |||
BX463485.2 | - | 5 | 468 | |||
BX951630.1 | - | 5 | 520 |