Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168222.1 | NP_001161694.1 | 10 | 1350 | ||
NM_024682.2 | NP_078958.2 | 11 | 1449 | |||
XM_011527317.2 | XP_011525619.1 | 11 | 1361 | |||
GenBank mRNA | AB449903.1 | - | 11 | 1150 | ||
AK022230.1 | - | 11 | 1208 | |||
AK090606.1 | - | 10 | 1150 | |||
AK300007.1 | - | 10 | 1109 | |||
AY129013.1 | - | 9 | 1896 | |||
BC003516.1 | - | 11 | 1302 | 3050014 |