Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBBX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199201.1 | NP_001186130.1 | 12 | 1219 | ||
NM_001199202.1 | NP_001186131.1 | 12 | 1167 | |||
NM_024687.3 | NP_078963.2 | 12 | 1219 | |||
XM_011513144.1 | XP_011511446.1 | 12 | 1219 | |||
XM_011513145.2 | XP_011511447.2 | 11 | 1245 | |||
XM_011513148.2 | XP_011511450.2 | 12 | 1219 | |||
XM_011513149.2 | XP_011511451.2 | 13 | 1336 | |||
XM_011513151.2 | XP_011511453.2 | 14 | 1525 | |||
XM_011513152.2 | XP_011511454.2 | 6 | 646 | |||
XM_011513153.2 | XP_011511455.2 | 3 | 318 | |||
XM_011513155.1 | XP_011511457.1 | 12 | 1219 | |||
XM_011513156.1 | XP_011511458.1 | 12 | 1219 | |||
XM_017007186.1 | XP_016862675.1 | 12 | 1219 | |||
XM_017007187.1 | XP_016862676.1 | 12 | 1219 | |||
XM_017007188.1 | XP_016862677.1 | 12 | 1219 | |||
XM_017007189.1 | XP_016862678.1 | 11 | 1245 | |||
XM_017007190.1 | XP_016862679.1 | 12 | 1219 | |||
GenBank mRNA | AK026702.1 | - | 2 | 68 | ||
AK093296.1 | - | 12 | 1205 | |||
BC034229.1 | - | 12 | 1167 | |||
BC144444.1 | - | 11 | 1166 | 9052969 |