Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 1486 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 1447 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 1444 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 1387 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 1348 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 1345 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 1384 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 1403 | |||
NM_024691.3 | NP_078967.3 | 5 | 1483 | |||
GenBank mRNA | AB209371.1 | - | 3 | 4732 | ||
AK026886.1 | - | 4 | 1357 | |||
AK302135.1 | - | 3 | 1306 | |||
AK304569.1 | - | 4 | 1392 | |||
AK307802.1 | - | 5 | 1475 | |||
AK309427.1 | - | 3 | 1950 | |||
AK316156.1 | - | 4 | 1405 | |||
AK316297.1 | - | 4 | 1437 | |||
BC008297.1 | - | 5 | 1429 | 3544044 | ||
BC024259.1 | - | 5 | 1429 | 3604215 |