Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098491.1 | NP_001091961.1 | 5 | 961 | ||
NM_001098492.1 | NP_001091962.1 | 4 | 922 | |||
NM_001098493.1 | NP_001091963.1 | 4 | 919 | |||
NM_001098494.1 | NP_001091964.1 | 4 | 862 | |||
NM_001098495.1 | NP_001091965.1 | 3 | 823 | |||
NM_001098496.1 | NP_001091966.1 | 3 | 820 | |||
NM_001291743.1 | NP_001278672.1 | 4 | 859 | |||
NM_001291744.1 | NP_001278673.1 | 4 | 878 | |||
NM_024691.3 | NP_078967.3 | 5 | 958 | |||
GenBank mRNA | AB209371.1 | - | 3 | 4207 | ||
AK026886.1 | - | 4 | 832 | |||
AK302135.1 | - | 3 | 781 | |||
AK304569.1 | - | 4 | 867 | |||
AK307802.1 | - | 5 | 950 | |||
AK309427.1 | - | 3 | 1425 | |||
AK316156.1 | - | 4 | 880 | |||
AK316297.1 | - | 4 | 912 | |||
BC008297.1 | - | 5 | 904 | 3544044 | ||
BC024259.1 | - | 5 | 904 | 3604215 |