Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031719.2 | NP_001026889.1 | 6 | 571 | ||
NM_001270424.1 | NP_001257353.1 | 7 | 654 | |||
NM_024705.2 | NP_078981.1 | 6 | 571 | |||
XM_005266527.3 | XP_005266584.1 | 6 | 577 | |||
XM_005266528.4 | XP_005266585.1 | 7 | 698 | |||
XM_011535235.2 | XP_011533537.1 | 8 | 767 | |||
XM_011535236.2 | XP_011533538.1 | 7 | 698 | |||
XM_011535237.1 | XP_011533539.1 | 7 | 646 | |||
XM_011535238.1 | XP_011533540.1 | 6 | 577 | |||
XM_017020749.1 | XP_016876238.1 | 8 | 767 | |||
XM_017020750.1 | XP_016876239.1 | 7 | 698 | |||
XM_017020751.1 | XP_016876240.1 | 6 | 577 | |||
XM_017020752.1 | XP_016876241.1 | 6 | 577 | |||
XM_017020753.1 | XP_016876242.1 | 5 | 515 | |||
XM_017020754.1 | XP_016876243.1 | 5 | 511 | |||
GenBank mRNA | AK023701.1 | - | 6 | 566 | ||
BC009825.2 | - | 6 | 507 | 3942115 | ||
BC026024.1 | - | 7 | 587 |