Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007269.1 | NP_001007270.1 | 2 | 1470 | ||
NM_001007270.1 | NP_001007271.1 | 3 | 1507 | |||
NM_024707.2 | NP_078983.1 | 3 | 1593 | |||
GenBank mRNA | AK024018.1 | - | 3 | 1577 |