Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007269.1 | NP_001007270.1 | 2 | 424 | ||
NM_001007270.1 | NP_001007271.1 | 3 | 461 | |||
NM_001319054.1 | NP_001305983.1 | 2 | 403 | |||
NM_001319055.1 | NP_001305984.1 | 2 | 450 | |||
NM_024707.2 | NP_078983.1 | 3 | 547 | |||
XM_005259262.3 | XP_005259319.1 | 3 | 675 | |||
XM_005259263.4 | XP_005259320.1 | 2 | 413 | |||
XM_017027312.1 | XP_016882801.1 | 2 | 650 | |||
GenBank mRNA | AK024018.1 | - | 3 | 531 | ||
AY114106.1 | - | 1 | 310 | |||
BC007793.2 | - | 3 | 519 | 4053402 | ||
BM009097.1 | - | 2 | 405 | |||
BU602037.1 | - | 3 | 550 | |||
CB148876.1 | - | 3 | 463 | |||
CR457321.1 | - | 1 | 310 |