Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277127.1 | NP_001264056.1 | 6 | 1046 | ||
NM_001277128.1 | NP_001264057.1 | 5 | 878 | |||
NM_024727.3 | NP_079003.2 | 6 | 1046 | |||
XM_011513158.2 | XP_011511460.1 | 6 | 1046 | |||
XM_011513160.2 | XP_011511462.1 | 6 | 1046 | |||
XM_017007204.1 | XP_016862693.1 | 6 | 767 | |||
GenBank mRNA | AK026912.1 | - | 5 | 874 | ||
AK097338.1 | - | 6 | 995 | |||
AY358138.1 | - | 6 | 976 | |||
BC117401.1 | - | 6 | 944 | 40125952 | ||
BC143569.1 | - | 6 | 945 | 9052080 | ||
BC143582.1 | - | 6 | 944 | |||
EU668335.1 | - | 6 | 995 |