Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077186.1 | NP_001070654.1 | 30 | 4038 | ||
NM_001145809.1 | NP_001139281.1 | 31 | 4137 | |||
NM_024729.3 | NP_079005.3 | 29 | 4014 | |||
XM_006723386.3 | XP_006723449.1 | 30 | 4099 | |||
XM_011527320.2 | XP_011525622.1 | 30 | 4116 | |||
XM_011527321.2 | XP_011525623.1 | 29 | 4092 | |||
XM_011527323.2 | XP_011525625.1 | 30 | 4106 | |||
GenBank mRNA | AB111886.1 | - | 20 | 2696 | ||
AB290169.1 | - | 31 | 4107 | |||
AY165122.1 | - | 30 | 4014 | |||
AY203926.1 | - | 8 | 1079 | |||
BC004396.1 | - | 2 | 81 | |||
BC129803.1 | - | 18 | 2111 |