Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282683.1 | NP_001269612.1 | 3, 4 | 216 | ||
NM_024735.4 | NP_079011.3 | 2, 3 | 468 | |||
GenBank mRNA | AF318348.1 | - | 1, 2 | 668 | ||
AF428140.1 | - | 2, 3 | 432 | |||
AK311122.1 | - | 2, 3 | 571 | |||
BC012748.1 | - | 2, 3 | 442 | 3958783 | ||
BG718342.1 | - | 3, 4 | 215 | |||
BI596608.1 | - | 2, 3 | 470 |