Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024738.2 | NP_079014.1 | 4 | 658 | ||
XM_005253937.3 | XP_005253994.1 | 3 | 495 | |||
GenBank mRNA | AK025068.1 | - | 4 | 626 | ||
AK222983.1 | - | 4 | 638 | |||
AK298857.1 | - | 3 | 500 | |||
AK309652.1 | - | 3 | 332 | |||
BC019843.1 | - | 4 | 518 | 5112874 | ||
BG537618.1 | - | 3 | 505 |