Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001895.3 | NP_001886.1 | 12 | 1277 | ||
NM_177559.2 | NP_808227.1 | 13 | 1394 | |||
NM_177560.2 | NP_808228.1 | 11 | 1067 | |||
GenBank mRNA | AB451279.1 | - | 11 | 1001 | ||
AB451411.1 | - | 11 | 1001 | |||
AK302583.1 | - | 11 | 956 | |||
AK312535.1 | - | 13 | 1285 | |||
BC011668.2 | - | 12 | 1149 | 4121882 | ||
BC050036.1 | - | 12 | 1168 | 5298655 | ||
BC053532.1 | - | 11 | 1096 | 3908058 | ||
BC071167.1 | - | 11 | 1008 | 5772615 | ||
BT019792.1 | - | 11 | 1001 | |||
DQ354583.1 | - | 12 | 1143 | |||
J02853.1 | - | 12 | 1154 | |||
M55265.1 | - | 12 | 1149 | |||
S53149.1 | - | 12 | 1154 |
Gene Symbol: CSNK2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256686.1 | NP_001243615.1 | 1 | 1239 |