Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324318.1 | NP_001311247.1 | 13 | 1682 | ||
NM_001324319.1 | NP_001311248.1 | 12 | 1562 | |||
NM_024745.4 | NP_079021.3 | 13 | 1793 | |||
NR_136738.1 | 14 | 1873 | ||||
GenBank mRNA | AK025662.1 | - | 9 | 1099 | ||
AK055931.1 | - | 13 | 1764 | |||
AK225315.1 | - | 9 | 1099 | |||
AK292164.1 | - | 13 | 1760 | |||
AK315706.1 | - | 13 | 1762 | |||
BC000960.1 | - | 8 | 877 | |||
BC030699.1 | - | 13 | 1768 | 4829528 |