Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324318.1 | NP_001311247.1 | 6 | 791 | ||
NM_024745.4 | NP_079021.3 | 6 | 902 | |||
NR_136738.1 | 6 | 905 | ||||
XM_011523335.2 | XP_011521637.1 | 6 | 4436 | |||
GenBank mRNA | AK025662.1 | - | 2 | 208 | ||
AK055931.1 | - | 6 | 873 | |||
AK225315.1 | - | 2 | 208 | |||
AK292164.1 | - | 6 | 869 | |||
AK310430.1 | - | 6 | 871 | |||
AK315706.1 | - | 6 | 871 | |||
BC030699.1 | - | 6 | 877 | 4829528 |