Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1010 | |||
NR_136193.1 | 4 | 1707 | ||||
NR_136194.1 | 4 | 1177 | ||||
NR_136197.1 | 4 | 1081 | ||||
NR_136198.1 | 3 | 898 | ||||
NR_136199.1 | 5 | 1220 | ||||
NR_136200.1 | 4 | 1710 | ||||
NR_136201.1 | 4 | 1877 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1618 | ||
AK021601.1 | - | 1 | 1357 | |||
AK300846.1 | - | 4 | 1088 | |||
BG519698.1 | - | 3 | 775 | |||
EU000853.1 | - | 4 | 923 | |||
EU000854.1 | - | 4 | 923 | |||
EU000855.1 | - | 3 | 2015 | |||
EU000856.1 | - | 3 | 2015 |