Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321635.1 | NP_001308564.1 | 4, 5 | 685 | ||
NM_001321636.1 | NP_001308565.1 | 4, 5 | 685 | |||
NM_024759.2 | NP_079035.1 | 4, 5 | 685 | |||
NR_135747.1 | 4, 5 | 685 | ||||
XM_011517302.2 | XP_011515604.1 | 4, 5 | 1719 | |||
GenBank mRNA | AK024017.1 | - | 4, 5 | 685 | ||
AK302414.1 | - | 2, 3 | 200 | |||
BC132687.1 | - | 4, 5 | 628 | 40146709 | ||
BC144055.1 | - | 4, 5 | 628 | 9052575 | ||
BP346328.1 | - | 3, 4 | 275 | |||
DA184376.1 | - | 4, 5 | 364 | |||
DC402113.1 | - | 2, 3 | 201 |