Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321635.1 | NP_001308564.1 | 10 | 1228 | ||
NM_024759.2 | NP_079035.1 | 10 | 1228 | |||
NR_135745.1 | 11 | 1392 | ||||
NR_135746.1 | 12 | 1514 | ||||
NR_135747.1 | 11 | 1452 | ||||
GenBank mRNA | AK024017.1 | - | 10 | 1228 | ||
AK302414.1 | - | 9 | 991 | |||
BC132687.1 | - | 10 | 1171 | 40146709 | ||
BC144055.1 | - | 10 | 1171 | 9052575 |