Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289088.1 | NP_001276017.1 | 11 | 2149 | ||
NM_001289089.1 | NP_001276018.1 | 10 | 1808 | |||
NM_001289090.1 | NP_001276019.1 | 11 | 1913 | |||
NM_001289091.1 | NP_001276020.1 | 11 | 1983 | |||
NM_001319955.1 | NP_001306884.1 | 9 | 1695 | |||
NM_024772.4 | NP_079048.3 | 10 | 1865 | |||
XM_011542163.2 | XP_011540465.1 | 9 | 1747 | |||
XM_011542165.2 | XP_011540467.1 | 11 | 1928 | |||
XM_011542166.2 | XP_011540468.1 | 11 | 1716 | |||
XM_017002358.1 | XP_016857847.1 | 8 | 1639 | |||
XM_017002359.1 | XP_016857848.1 | 9 | 1656 | |||
XM_017002360.1 | XP_016857849.1 | 11 | 1922 | |||
XM_017002361.1 | XP_016857850.1 | 4 | 778 | |||
GenBank mRNA | AK096206.1 | - | 11 | 1961 | ||
AK299779.1 | - | 10 | 1808 | |||
BX537537.1 | - | 10 | 1837 | |||
BX648307.1 | - | 9 | 1668 |