Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145348.1 | NP_001138820.1 | 4 | 1019 | ||
NM_024773.2 | NP_079049.2 | 4 | 969 | |||
XM_017023676.1 | XP_016879165.1 | 4 | 969 | |||
XM_017023677.1 | XP_016879166.1 | 5 | 1054 | |||
XM_017023678.1 | XP_016879167.1 | 5 | 1050 | |||
XM_017023679.1 | XP_016879168.1 | 5 | 1050 | |||
XM_017023680.1 | XP_016879169.1 | 2 | 561 | |||
GenBank mRNA | AK023860.1 | - | 4 | 960 | ||
AK225066.1 | - | 4 | 926 | |||
AK297166.1 | - | 4 | 1019 | |||
AK298410.1 | - | 1 | 89 | |||
AK308499.1 | - | 2 | 283 | |||
BC027911.1 | - | 4 | 948 | 5207043 | ||
KU178785.1 | - | 3 | 787 | |||
KU178786.1 | - | 3 | 787 |