Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076786.2 | NP_001070254.1 | 4 | 1481 | ||
XM_006718325.3 | XP_006718388.3 | 4 | 1833 | |||
XM_017018327.1 | XP_016873816.1 | 4 | 1833 | |||
XM_017018328.1 | XP_016873817.1 | 4 | 1833 | |||
XM_017018329.1 | XP_016873818.1 | 2 | 2286 | |||
XM_017018330.1 | XP_016873819.1 | 3 | 1231 |