Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024775.9 | NP_079051.9 | 2 | 217 | ||
GenBank mRNA | AF453443.1 | - | 1 | 101 | ||
AK027112.1 | - | 2 | 187 | |||
AK315627.1 | - | 2 | 196 | |||
BC018195.1 | - | 2 | 158 | 3863118 | ||
BC029584.1 | - | 4 | 666 | |||
BM471707.1 | - | 2 | 163 |